Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ppn
Entrez Gene ID
43872
Gene Name
Papilin
Gene Aliases
CG1540, CG18436, CG33103, CT3186, CT41988, Dmel\CG33103, Dmel_CG33103, ppl, ppn
Chromosome Location
Chr.3R: 28520665 - 28540324 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003137
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170290.2 NP_001163761.1 10 - 11 7157 84
NM_176575.3 NP_788752.2 11 - 12 7328 84

View Full Product Details