Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Myo95E
Entrez Gene ID
2768680
Gene Name
Myosin 95E
Gene Aliases
95E, CG31134, CG31135, CG5501, Dm95E, DmAE003746, Dmel\CG5501, Dmel_CG5501, Myo5, myo 95E
Chromosome Location
Chr.3R: 24154826 - 24161498 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0039157
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001032033.2 NP_001027204.2 2 - 3 252 67
NM_001032035.2 NP_001027206.1 1 - 2 355 67
NM_001032036.2 NP_001027207.1 2 - 3 252 67
NM_001032037.2 NP_001027208.1 1 - 2 355 67
NM_206555.2 NP_996278.1 1 - 2 355 67
NM_206556.2 NP_996279.1 1 - 2 355 67
GenBank mRNA AF454350.1 - 1 - 2 355 67
AF454351.1 - 1 - 2 355 67
BT011452.1 - 2 - 3 249 67

View Full Product Details