Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Myo95E
Entrez Gene ID
2768680
Gene Name
Myosin 95E
Gene Aliases
95E, CG31134, CG31135, CG5501, Dm95E, DmAE003746, Dmel\CG5501, Dmel_CG5501, Myo5, myo 95E
Chromosome Location
Chr.3R: 24154826 - 24161498 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0039157
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001032033.2 NP_001027204.2 6 - 7 1547 113
NM_001032035.2 NP_001027206.1 5 - 6 1666 113
NM_001032036.2 NP_001027207.1 6 - 7 1563 113
NM_001032037.2 NP_001027208.1 6 - 7 1680 113
NM_206555.2 NP_996278.1 5 - 6 1650 113
NM_206556.2 NP_996279.1 5 - 6 1650 113
GenBank mRNA AF454350.1 - 5 - 6 1650 113
AF454351.1 - 5 - 6 1650 113
BT011452.1 - 6 - 7 1560 113

View Full Product Details