Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- amd
- Entrez Gene ID
-
35188
- Gene Name
- alpha methyl dopa-resistant
- Gene Aliases
-
AMD-r, CG10501, CG17345, Dmel\CG10501, Dmel_CG10501, X04695, fAMD, l(2)37Bk, l(2)amd, l(2)amd alpha-methyldopa hypersensitive, l(2)amd[H]
- Chromosome Location
-
Chr.2L: 19110154 - 19114110 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0000075
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057244.4 |
NP_476592.1 |
1 - 2 |
473 |
110 |
GenBank mRNA |
BT133174.1 |
- |
1 - 2 |
469 |
110 |
View Full Product Details