Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CecB
- Entrez Gene ID
-
43598
- Gene Name
- Cecropin B
- Gene Aliases
-
CG1878, Cec, Cec B, Dmel\CG1878, Dmel_CG1878, cec, cecB, cedcB, crpB
- Chromosome Location
-
Chr.3R: 30213315 - 30213797 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0000278
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079851.3 |
NP_524590.1 |
1 - 2 |
177 |
91 |
View Full Product Details