Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Ubx
- Entrez Gene ID
-
42034
- Gene Name
- Ultrabithorax
- Gene Aliases
-
BX-C, Bxl, CG10388, Cbx, DUbx, Dm Ubx, DmUbx, Dmel\CG10388, Dmel_CG10388, Hm, UBX, UBx, Uab, Ubx1b, abx, bx, bx[D], bxd, bxd[D], bxl, l(3)89Eb, pbx, ubx
- Chromosome Location
-
Chr.3R: 16656623 - 16734426 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0003944
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_080500.4 |
NP_536748.1 |
1 - 2 |
1705 |
73 |
View Full Product Details