Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
spi
Entrez Gene ID
35253
Gene Name
spitz
Gene Aliases
CG10334, CT29014, Dmel\CG10334, Dmel_CG10334, EP(2)2378, Spi, Spitz, anon-WO0118547.158, l(2)01068, l(2)37Fa, l(2)s3547, s-spi, sSpi
Chromosome Location
Chr.2L: 19567900 - 19577315 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0005672
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001032110.2 NP_001027281.1 3 - 3 1253 99
NM_057561.6 NP_476909.2 2 - 2 1134 99
NM_134291.5 NP_599118.2 3 - 3 1330 99
NM_134292.3 NP_599119.2 2 - 2 1115 99
NM_134293.3 NP_599120.2 3 - 3 1311 99
GenBank mRNA BT044503.1 - 2 - 2 1144 99

View Full Product Details