Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
caps
Entrez Gene ID
39493
Gene Name
capricious
Gene Aliases
BcDNA:LD07388, CAPS, CG11282, CT31495, Caps, Dmel\CG11282, Dmel_CG11282, l(3)02937, l(3)rM632
Chromosome Location
Chr.3L: 13228692 - 13279642 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0023095
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001043139.4 NP_001036604.2 5 - 5 2638 64
NM_001316462.1 NP_001303391.1 5 - 5 2638 64
NM_079332.4 NP_524056.3 5 - 5 2638 64
NM_168540.3 NP_729880.2 4 - 4 2425 64
GenBank mRNA BT016006.1 - 4 - 4 2423 64

View Full Product Details