Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Eig71Ee
- Entrez Gene ID
-
39706
- Gene Name
- Ecdysone-induced gene 71Ee
- Gene Aliases
-
CG7604, Dmel\CG7604, Dmel_CG7604, I71-7, L71, L71-7, VII, eig71Ee, gene VII, gene-VII, gp150
- Chromosome Location
-
Chr.3L: 15654472 - 15656010 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0004592
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079368.3 |
NP_524092.2 |
2 - 2 |
1300 |
87 |
GenBank mRNA |
AY119006.1 |
- |
2 - 2 |
1288 |
87 |
View Full Product Details