Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Act88F
- Entrez Gene ID
-
41885
- Gene Name
- Actin 88F
- Gene Aliases
-
Act(88F), Act88-F, Act88f, Actin, Actin88F, ArpD, CG5178, Dmel\CG5178, Dmel_CG5178, E, Ifm(3)7, P83967, act 88F, act88F, actin, rsd
- Chromosome Location
-
Chr.3R: 15439969 - 15442177 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0000047
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079643.2 |
NP_524367.1 |
3 - 3 |
1483 |
123 |
GenBank mRNA |
BT050557.1 |
- |
3 - 3 |
1485 |
123 |
View Full Product Details