Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- mRpL12
- Entrez Gene ID
-
44326
- Gene Name
- mitochondrial ribosomal protein L12
- Gene Aliases
-
BcDNA:RE53508, CG5012, Dmel\CG5012, Dmel_CG5012, L7, L7/12A, L7/L12, Mrp17, l(3)10534, l(3)j4B2, l(3)rL424, mRpL7-L12
- Chromosome Location
-
Chr.3L: 8956791 - 8957584 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0011787
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_080003.4 |
NP_524742.1 |
2 - 2 |
657 |
63 |
GenBank mRNA |
AY060871.1 |
- |
2 - 2 |
599 |
63 |
View Full Product Details