Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Pp1-13C
- Entrez Gene ID
-
48531
- Gene Name
- Protein phosphatase 1 at 13C
- Gene Aliases
-
13C, CG9156, DmPp1-13C, Dmel\CG9156, Dmel_CG9156, PP1, PP1 13C, PP1 beta13c, PP1-13C, PP13C, PP1alpha-13C, PP1alpha13C, PP1c, Pp1, pp1-13C
- Chromosome Location
-
Chr.X: 15359101 - 15361061 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0003132
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_080182.3 |
NP_524921.1 |
2 - 2 |
1761 |
96 |
GenBank mRNA |
AY060272.1 |
- |
2 - 2 |
1761 |
96 |
View Full Product Details