Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Phk-3
Entrez Gene ID
37997
Gene Name
Pherokine 3
Gene Aliases
BcDNA:RE09339, CG9358, DmelCSP3, Dmel\CG9358, Dmel_CG9358, PHK-3
Chromosome Location
Chr.2R: 24970843 - 24973207 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0035089
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299941.1 NP_001286870.1 2 - 2 559 76
NM_001299942.1 NP_001286871.1 2 - 2 642 76
NM_138146.3 NP_611990.1 2 - 2 470 76
GenBank mRNA AY071015.1 - 2 - 2 467 76
BT120076.1 - 2 - 2 532 76

View Full Product Details