Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CheA75a
- Entrez Gene ID
-
40011
- Gene Name
- Chemosensory protein A 75a
- Gene Aliases
-
BcDNA:HL02449, CG7313, Dmel\CG7313, Dmel_CG7313
- Chromosome Location
-
Chr.3L: 18252088 - 18252846 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0036783
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_140778.2 |
NP_649035.2 |
2 - 2 |
560 |
92 |
View Full Product Details