Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Nplp3
- Entrez Gene ID
-
59235
- Gene Name
- Neuropeptide-like precursor 3
- Gene Aliases
-
BcDNA:RH16169, CG13061, CG18502, Dmel\CG13061, Dmel_CG13061, NPLP3, NpIp3, SHA, VVIamide, nplp3
- Chromosome Location
-
Chr.3L: 16318024 - 16318690 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0042201
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_144453.3 |
NP_652710.1 |
2 - 2 |
448 |
124 |
GenBank mRNA |
AY113570.1 |
- |
2 - 2 |
447 |
124 |
View Full Product Details