Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ubi-p5E
Entrez Gene ID
326237
Gene Name
Ubiquitin-5E
Gene Aliases
CG18282, CG32744, CR32744, DmUb, DmUbi-p5E, Dmel\CG32744, Dmel_CG32744, UB, Ub, Ubi, Ubiq
Chromosome Location
Chr.X: 6276876 - 6279647 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0086558
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298008.1 NP_001284937.1 2 - 2 2112 95
NM_167065.3 NP_727078.1 2 - 2 1739 95
GenBank mRNA BT044587.1 - 2 - 2 1722 95

View Full Product Details