Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pfrx
Entrez Gene ID
32938
Gene Name
6-phosphofructo-2-kinase
Gene Aliases
CG3400, Dmel\CG3400, Dmel_CG3400, PFK2, Pfk2
Chromosome Location
Chr.X: 19495505 - 19509350 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0027621
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258821.2 NP_001245750.1 3 - 3 1725 64
NM_001258822.2 NP_001245751.1 2 - 2 1861 64
NM_001258823.2 NP_001245752.1 2 - 2 2047 64
NM_001298511.1 NP_001285440.1 2 - 2 1695 64
NM_001298512.1 NP_001285441.1 2 - 2 2037 64
NM_058103.3 NP_477451.1 2 - 2 2040 64
NM_058104.5 NP_477452.3 2 - 2 2040 64
NM_167651.2 NP_728249.1 2 - 2 1645 64
NM_167652.2 NP_728250.1 2 - 2 1647 64
NM_167653.3 NP_728251.1 2 - 2 1857 64
NM_167654.2 NP_728252.1 2 - 2 1576 64
NM_167655.2 NP_728253.1 2 - 2 1644 64
NM_167656.2 NP_728254.1 2 - 2 1642 64
GenBank mRNA AB023511.1 - 2 - 2 2040 64
AY122115.1 - 2 - 2 2042 64

View Full Product Details