Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SIFa
Entrez Gene ID
3346202
Gene Name
SIFamide
Gene Aliases
CG33527, Dmel\CG33527, Dmel_CG33527, Drm-IFa, Drm-SIFa, IFAMIDE, IFa, Neb-LFamide, SIFR
Chromosome Location
Chr.2R: 24577566 - 24578143 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0053527
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259567.2 NP_001246496.1 2 - 2 400 113
GenBank mRNA BT030951.1 - - 515 113

View Full Product Details