Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nab
Entrez Gene ID
3346237
Gene Name
CG33545-PB
Gene Aliases
CG15000, CG15001, CG33545, Dmel\CG33545, Dmel_CG33545, NAB, Nab, dNAB, dnab, l(2)SH3 143, l(3)64Aj, l(3)SH143, l(3)SH21
Chromosome Location
Chr.3L: 4153618 - 4160932 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0259986
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014557.2 NP_001014557.3 1 - 2 603 71
NM_001169873.1 NP_001163344.1 1 - 2 603 71
NM_001274478.1 NP_001261407.1 1 - 2 603 71

View Full Product Details