Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Ssl
- Entrez Gene ID
-
37909
- Gene Name
- Suppressor of Stellate-like
- Gene Aliases
-
CG13591, CK2betates, CKIIb1, Dm-CK2-beta[Tes], DmCK2betates, Dmel\CG13591, Dmel_CG13591, NEST:bs30a09, SSL, Su(Ste)l, betaCK2tes
- Chromosome Location
-
Chr.2R: 24499243 - 24500267 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0015300
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079124.3 |
NP_523848.2 |
3 - 3 |
674 |
53 |
GenBank mRNA |
AY089437.1 |
- |
3 - 3 |
628 |
53 |
View Full Product Details