Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RhoGAP1A
Entrez Gene ID
5740235
Gene Name
Rho GTPase activating protein at 1A
Gene Aliases
BCR, CG17617, CG17960, CG40453, CG40494, Dmel\CG40494, Dmel_CG40494, EG:23E12.2, EG:23E12.5, RhoGAP, rhoGAP1A
Chromosome Location
Chr.X: 127449 - 140340 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0025836
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103386.2 NP_001096856.1 14 - 15 4524 73

View Full Product Details