Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- nompB
- Entrez Gene ID
-
35414
- Gene Name
- no mechanoreceptor potential B
- Gene Aliases
-
146590_s_at, CG12548, CG14465, CG15190, CG4137, Dmel\CG12548, Dmel_CG12548, IFT88, NOMPB, NompB, OSM-5, Tg737, nomp, osm-5
- Chromosome Location
-
Chr.2L: 21578292 - 21589423 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0016919
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_165385.4 |
NP_724347.3 |
3 - 4 |
619 |
79 |
View Full Product Details