Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CheB93b
- Entrez Gene ID
-
318736
- Gene Name
- Chemosensory protein B 93b
- Gene Aliases
-
CG31438, Dmel\CG31438, Dmel_CG31438
- Chromosome Location
-
Chr.3R: 21409123 - 21409924 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0051438
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_169959.3 |
NP_732641.2 |
2 - 3 |
447 |
99 |
View Full Product Details