Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ak1
Entrez Gene ID
445486
Gene Name
adenylate kinase 1
Gene Aliases
zgc:91930
Chromosome Location
Chr.5: 71699874 - 71724814 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003993.1 NP_001003993.1 6 - 7 693 117
XM_005168682.4 XP_005168739.1 5 - 6 590 117
XM_005168683.4 XP_005168740.1 6 - 7 774 117
XM_009302056.3 XP_009300331.1 6 - 7 690 117
XM_017356070.2 XP_017211559.1 7 - 8 903 117
GenBank mRNA BC080261.1 - 6 - 7 693 117

View Full Product Details