Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
eef2kmt
Entrez Gene ID
450038
Gene Name
eukaryotic elongation factor 2 lysine methyltransferase
Gene Aliases
fam86a, zgc:101751
Chromosome Location
Chr.3: 28869610 - 28872929 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006058.1 NP_001006058.1 6 - 7 792 81
XM_009299451.2 XP_009297726.1 6 - 7 918 81
GenBank mRNA BC083266.1 - 6 - 7 792 81
BC090275.1 - 1 - 2 1040 81

View Full Product Details