Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nsfb
Entrez Gene ID
554200
Gene Name
N-ethylmaleimide-sensitive factor b
Gene Aliases
-
Chromosome Location
Chr.12: 248424 - 299403 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024454.2 NP_001019625.2 18 - 19 2093 86
NM_001354163.1 NP_001341092.1 18 - 19 2093 86
XM_005156207.4 XP_005156264.1 18 - 19 2130 86
XM_017358461.2 XP_017213950.1 17 - 18 1940 86
GenBank mRNA BC162793.1 - 18 - 19 2071 86
BC162811.1 - 18 - 19 2071 86
DQ022212.1 - 18 - 19 2093 86

View Full Product Details