Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
elk3
Entrez Gene ID
557074
Gene Name
ETS transcription factor ELK3
Gene Aliases
si:ch211-172b12.2, si:dkey-201e17.1, zgc:198153, zgc:198183
Chromosome Location
Chr.4: 259508 - 273601 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030108.1 NP_001025279.1 1 - 2 213 109
XM_005164542.4 XP_005164599.1 2 - 3 585 109
GenBank mRNA BC171426.1 - 1 - 2 275 109
BC171456.1 - 1 - 2 275 109

View Full Product Details