Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
mib1
Entrez Gene ID
352910
Gene Name
mindbomb E3 ubiquitin protein ligase 1
Gene Aliases
cg5841, chunp6889, fe47f05, im:7148100, mib, wu:fe47f05
Chromosome Location
Chr.2: 4074171 - 4161272 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173286.3 NP_775393.2 10 - 11 1575 64
XM_009296844.3 XP_009295119.1 10 - 11 2510 64
XM_009296845.3 XP_009295120.1 10 - 11 2510 64
XM_009296846.3 XP_009295121.1 10 - 11 2510 64
XM_009296848.3 XP_009295123.1 10 - 11 2510 64
XM_009296854.3 XP_009295129.1 10 - 11 2477 64
XM_009296859.3 XP_009295134.1 10 - 11 2510 64
GenBank mRNA AF537301.1 - 10 - 11 1575 64
BC162476.1 - 10 - 11 1568 64

View Full Product Details