Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fam49ba
Entrez Gene ID
337591
Gene Name
family with sequence similarity 49 member Ba
Gene Aliases
fam49b, fj85c11, wu:fj85c11, zgc:64118
Chromosome Location
Chr.2: 32215272 - 32262287 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291713.1 NP_001278642.1 8 - 9 1092 77
NM_200070.3 NP_956364.2 10 - 11 1299 77
GenBank mRNA AY394961.1 - 10 - 11 999 77
BC053261.1 - 8 - 9 778 77
DT055468.1 - 1 - 2 483 77

View Full Product Details