Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pygmb
Entrez Gene ID
393444
Gene Name
phosphorylase, glycogen, muscle b
Gene Aliases
zgc:63642
Chromosome Location
Chr.7: 22823889 - 22852174 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001328031.1 NP_001314960.1 20 - 21 2701 79
GenBank mRNA BC055181.1 - 9 - 10 1120 79

View Full Product Details