Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sra1
Entrez Gene ID
415137
Gene Name
steroid receptor RNA activator 1
Gene Aliases
zgc:86613
Chromosome Location
Chr.21: 43882274 - 43891419 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002047.1 NP_001002047.1 2 - 3 215 61
GenBank mRNA BC071314.1 - 2 - 3 215 61

View Full Product Details