Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mrrf
Entrez Gene ID
431721
Gene Name
mitochondrial ribosome recycling factor
Gene Aliases
zgc:91914
Chromosome Location
Chr.10: 92473 - 133716 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002174.1 NP_001002174.1 4 - 5 453 60
GenBank mRNA BC074048.1 - 4 - 5 453 60

View Full Product Details