Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
crygn2
Entrez Gene ID
445034
Gene Name
crystallin, gamma N2
Gene Aliases
crygnb, zgc:92717
Chromosome Location
Chr.24: 41305391 - 41312451 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003428.3 NP_001003428.1 2 - 3 336 81
GenBank mRNA AY738746.1 - 2 - 3 310 81
BC076197.1 - 2 - 3 319 81
EH590493.1 - 2 - 3 348 81

View Full Product Details