Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
psmb1
Entrez Gene ID
445413
Gene Name
proteasome 20S subunit beta 1
Gene Aliases
wu:fc51f06, zgc:103665
Chromosome Location
Chr.13: 1142725 - 1151345 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003889.1 NP_001003889.1 4 - 5 468 86
GenBank mRNA AY648806.1 - 4 - 5 468 86
BC085580.1 - 4 - 5 463 86
BC165077.1 - 4 - 5 458 86

View Full Product Details