Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ech1
Entrez Gene ID
450048
Gene Name
enoyl CoA hydratase 1, peroxisomal
Gene Aliases
zgc:101710
Chromosome Location
Chr.15: 13436 - 32066 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006068.1 NP_001006068.1 6 - 7 741 96
GenBank mRNA BC083254.1 - 6 - 7 741 96
BC152159.1 - 6 - 7 541 96
BC154279.1 - 6 - 7 743 96
BC164085.1 - 6 - 7 559 96

View Full Product Details