Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cyp1d1
Entrez Gene ID
492344
Gene Name
cytochrome P450, family 1, subfamily D, polypeptide 1
Gene Aliases
zgc:92205
Chromosome Location
Chr.5: 25606658 - 25621320 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007310.1 NP_001007311.1 3 - 4 1072 151
GenBank mRNA BC085648.1 - 3 - 4 1072 151
FJ416496.1 - 3 - 4 1068 151

View Full Product Details