Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cyp2n13
Entrez Gene ID
492484
Gene Name
cytochrome P450, family 2, subfamily N, polypeptide 13
Gene Aliases
cyp2j30, si:dkey-183n20.13, zgc:103711
Chromosome Location
Chr.20: 25515259 - 25522781 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007356.1 NP_001007357.1 2 - 3 378 86
GenBank mRNA BC085592.1 - 2 - 3 378 86
BC164549.1 - 2 - 3 394 86

View Full Product Details