Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
map4k2
Entrez Gene ID
678610
Gene Name
mitogen-activated protein kinase kinase kinase kinase 2
Gene Aliases
dZ150L22.2, map4k2l, map4k2l-2, map4k3l, si:dz150l22.2, si:dz263j20.1, zgc:136670
Chromosome Location
Chr.7: 17648931 - 17690869 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040358.1 NP_001035448.1 12 - 13 1014 89
GenBank mRNA BC115221.1 - 12 - 13 1014 89
BC162619.1 - 12 - 13 1112 89

View Full Product Details