Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
btbd11a
Entrez Gene ID
562893
Gene Name
BTB (POZ) domain containing 11a
Gene Aliases
btbd11, si:ch211-161f1.1
Chromosome Location
Chr.4: 11813423 - 12007387 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045002.1 NP_001038467.1 9 - 10 1262 90
NM_001327842.1 NP_001314771.1 10 - 11 2471 90
XM_005164650.4 XP_005164707.1 9 - 10 2093 90
XM_009300161.2 XP_009298436.1 9 - 10 1259 90

View Full Product Details