Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
otomp
Entrez Gene ID
565818
Gene Name
otolith matrix protein
Gene Aliases
im:6904989, omp, omp-1, wu:fc44d11, zomp1
Chromosome Location
Chr.2: 41504752 - 41518340 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045087.1 NP_001038552.1 5 - 6 684 77
GenBank mRNA AY681353.1 - 5 - 6 684 77
BC076449.1 - 5 - 6 663 77
BC134025.1 - 5 - 6 716 77

View Full Product Details