Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
otomp
Entrez Gene ID
565818
Gene Name
otolith matrix protein
Gene Aliases
im:6904989, omp, omp-1, wu:fc44d11, zomp1
Chromosome Location
Chr.2: 41504752 - 41518340 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045087.1 NP_001038552.1 8 - 9 931 98
GenBank mRNA AY681353.1 - 8 - 9 931 98
BC076449.1 - 7 - 8 910 98
BC134025.1 - 7 - 8 963 98

View Full Product Details