Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dph7
Entrez Gene ID
564032
Gene Name
diphthamide biosynthesis 7
Gene Aliases
wdr85, zgc:162277
Chromosome Location
Chr.5: 25077212 - 25084347 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082839.1 NP_001076308.1 6 - 7 701 68
XM_005165174.4 XP_005165231.1 6 - 7 985 68
XM_009301350.3 XP_009299625.1 7 - 8 1004 68
XM_021476262.1 XP_021331937.1 6 - 7 985 68
XM_021476263.1 XP_021331938.1 6 - 7 988 68
GenBank mRNA BC133930.1 - 6 - 7 701 68

View Full Product Details