Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
il15ra
Entrez Gene ID
100192216
Gene Name
interleukin 15 receptor subunit alpha
Gene Aliases
il-15ra, zgc:195175
Chromosome Location
Chr.4: 32525 - 50390 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083584.2 NP_001077053.1 3 - 4 326 83
NM_001359927.1 NP_001346856.1 3 - 4 326 83
NM_001359929.1 NP_001346858.1 3 - 4 326 83
XM_005164796.3 XP_005164853.1 3 - 4 403 83
GenBank mRNA EF143578.1 - 3 - 4 326 83
EF165722.1 - 3 - 4 326 83
EF165724.1 - 3 - 4 326 83

View Full Product Details