Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ednrba
Entrez Gene ID
30442
Gene Name
endothelin receptor Ba
Gene Aliases
ednrb1, ednrb1a
Chromosome Location
Chr.1: 28624186 - 28629471 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131197.1 NP_571272.1 2 - 3 673 62
GenBank mRNA AF275636.1 - 2 - 3 673 62

View Full Product Details