Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
meis3
Entrez Gene ID
81885
Gene Name
myeloid ecotropic viral integration site 3
Gene Aliases
NH10, fc22g05, id:ibd3530, meis3.1, wu:fc22g05
Chromosome Location
Chr.15: 6838071 - 6863317 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131778.2 NP_571853.1 5 - 6 738 148
GenBank mRNA AF222995.1 - 5 - 6 566 148
BC075739.1 - 5 - 6 742 148

View Full Product Details