Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
terfa
Entrez Gene ID
192316
Gene Name
telomeric repeat binding factor a
Gene Aliases
CHUNP6903, wu:fi15d05, wu:fi22b03, zgc:111909, zgc:114011
Chromosome Location
Chr.4: 77973874 - 77996958 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173243.2 NP_775350.2 10 - 11 1457 78
XM_009301103.3 XP_009299378.1 10 - 11 1588 78
XM_009301104.3 XP_009299379.1 10 - 11 1586 78
XM_009301105.3 XP_009299380.1 9 - 10 1478 78
GenBank mRNA AY099522.1 - 13 - 14 1662 78
BC096857.1 - 9 - 10 1281 78
BC098889.1 - 9 - 10 1299 78
BC165460.1 - 9 - 10 1303 78

View Full Product Details