Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mylipa
Entrez Gene ID
335888
Gene Name
myosin regulatory light chain interacting protein a
Gene Aliases
Mir, mylip, wu:fj36b03, zgc:55987
Chromosome Location
Chr.19: 26340737 - 26378655 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199983.1 NP_956277.1 1 - 2 236 108
GenBank mRNA AY423020.1 - 1 - 2 236 108
AY434450.1 - 1 - 2 198 108
BC045844.1 - 1 - 2 224 108

View Full Product Details