Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
slc46a1
Entrez Gene ID
393255
Gene Name
solute carrier family 46 member 1
Gene Aliases
zgc:56400
Chromosome Location
Chr.15: 28175658 - 28186529 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200285.1 NP_956579.1 3 - 4 1240 95
GenBank mRNA BC049421.1 - 3 - 4 1240 95

View Full Product Details