Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mtmr8
Entrez Gene ID
393365
Gene Name
myotubularin related protein 8
Gene Aliases
si:dkey-286f3.1, zgc:56405
Chromosome Location
Chr.5: 20668 - 45134 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200394.1 NP_956688.1 8 - 9 1120 107
XM_021476036.1 XP_021331711.1 3 - 4 107
GenBank mRNA AY423041.1 - 8 - 9 107
BC053411.1 - 8 - 9 1120 107

View Full Product Details