Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pah
Entrez Gene ID
378962
Gene Name
phenylalanine hydroxylase
Gene Aliases
cb877, wu:fb02f10, zgc:65981
Chromosome Location
Chr.4: 17391680 - 17409367 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200551.1 NP_956845.1 12 - 13 1342 66
GenBank mRNA BC056537.1 - 12 - 13 1342 66

View Full Product Details